Learn More
Aviva Systems Biology Corp RIPK3 Antibody - N-terminal region (ARP32835_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32835_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RIPK3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RIPK3. Alternate names: RIP3ApplicationWB. Species reactivity: Cow, Horse, Human, Mouse, Rat
Catalog No. 50-014-54784