Learn More
Medchemexpress LLC Ripk3-in-1 | 2361139-70-8 | 99.3% | 50 MG
SDP
Catalog No. 5000341056
Encompass_Preferred
RIPK3-IN-1 is a type II DFG-out inhibitor with primary activity against RIPK3. It also demonstrates inhibitory effects on RIPK1, RIPK2, and c-Met kinase. Additionally, it inhibits ABL, BRAF/V599E, MAP4K3, and SRC kinases.
  • Inhibits RIPK3 with an IC50 of 9.1 nM
  • Inhibits RIPK1 with an IC50 of 5.5 μM
  • Inhibits RIPK2 with an IC50 of >10 μM
  • Inhibits c-Met kinase with an IC50 of 1.1 μM
  • Inhibits ABL with an IC50 of 0.37 μM
  • Inhibits BRAF/V599E with an IC50 of 0.15 μM
  • Inhibits MAP4K3 with an IC50 of 0.012 μM
  • Inhibits SRC with an IC50 of 0.075 μM
Catalog No. 50-003-41056 Supplier Medchemexpress LLC Supplier No. HY13106450MG
RIPK3-IN-1 is a type II DFG-out inhibitor with primary activity against RIPK3. It also demonstrates inhibitory effects on RIPK1, RIPK2, and c-Met kinase. Additionally, it inhibits ABL, BRAF/V599E, MAP4K3, and SRC kinases.
  • Inhibits RIPK3 with an IC50 of 9.1 nM
  • Inhibits RIPK1 with an IC50 of 5.5 μM
  • Inhibits RIPK2 with an IC50 of >10 μM
  • Inhibits c-Met kinase with an IC50 of 1.1 μM
  • Inhibits ABL with an IC50 of 0.37 μM
  • Inhibits BRAF/V599E with an IC50 of 0.15 μM
  • Inhibits MAP4K3 with an IC50 of 0.012 μM
  • Inhibits SRC with an IC50 of 0.075 μM