Learn More
Medchemexpress LLC Ripk3-in-1 | 2361139-70-8 | 99.3% | C29H25FN4O4 | 10MG
SDP
Catalog No. 5000258947
Encompass_Preferred
RIPK3-IN-1 is a small-molecule, type II DFG-out inhibitor of receptor-interacting protein kinase 3 (RIPK3) intended for research use in biochemical and cellular studies. It exhibits potent biochemical activity and is supplied as a high-purity compound suitable for in vitro investigations into necroptosis and related signaling pathways.
  • Potent RIPK3 inhibition, IC50 = 9.1 nM.
  • High purity, 99.3% by supplier analysis.
  • Chemical formula C29H25FN4O4 and molecular weight 512.53 g/mol.
  • Suitable for biochemical and cellular assays targeting necroptosis pathways.
  • Available in multiple sizes to support assay development and scale-up.
Catalog No. 50-002-58947 Supplier Medchemexpress LLC Supplier No. HY13106410MG
RIPK3-IN-1 is a small-molecule, type II DFG-out inhibitor of receptor-interacting protein kinase 3 (RIPK3) intended for research use in biochemical and cellular studies. It exhibits potent biochemical activity and is supplied as a high-purity compound suitable for in vitro investigations into necroptosis and related signaling pathways.
  • Potent RIPK3 inhibition, IC50 = 9.1 nM.
  • High purity, 99.3% by supplier analysis.
  • Chemical formula C29H25FN4O4 and molecular weight 512.53 g/mol.
  • Suitable for biochemical and cellular assays targeting necroptosis pathways.
  • Available in multiple sizes to support assay development and scale-up.