Learn More
Aviva Systems Biology Corp RIPK5 Antibody - middle region (ARP55938_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55938_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RIPK5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RIPK5. Alternate names: DustyPK, HDCMD38P, KIAA0472, RIP5, RIPK5ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61669