Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Supertek™ Ripple Tank
GREENER_CHOICESCI_ED

Catalog No. S139438 Shop All Supertek Scientific Products
Change view
Click to view available options
Grade:
9+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S139438 9+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S139438 Supplier Supertek™ Supplier No. PH36044
Add to Cart
Add to Cart
This item is not returnable. View return policy

Demonstrate all types of waves and wave motions using this quality Supertek™ Ripple Tank with accessories.

  • A constructive and functional way to demonstrate all types of waves and wave motion when using this 23 x 20 x 3 in. ripple tank supported on 4 legs with leveling feet.
  • The tank edges are shaped to minimize undesired reflections.
  • Tank has a drain hole closed by a rubber stopper.
  • Two light alloy uprights to carry a horizontal rippler support rod which is adjustable for height.
  • The ripple assembly elastically suspended from the support rod consists of a 4-6 V DC motor mounted on a wooden beam.
  • The motor has an eccentric weight which causes the beam to oscillate.
  • Two point sources are provided by two plastic spheres each mounted on a right angle rod.
  • A metal rod is fixed centrally into a socket in one short side of the frame to afford support to the illuminant.

Specifications

Grade 9+
Format Class Set
Includes 2 straight obstacles of aluminum alloy (5.12 in.), 1 straight obstacle (1.57 in.), 1 curved reflector (7.87 in. radius), 1 trapezoidal plastic plate for refraction, 1 wood hand wave roller, 1 water dropper, 1 Pk. of rubber bands, 1 foam plastic sponge, 1 illuminant with a 12 V, 24 W lamp (mounted in a lamp house and provided with a long supporting rod that can be attached to a retort stand), and Instruction manual.
Languages English
Subject Physical Science - Waves
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.