Learn More
Medchemexpress LLC Ripretinib | 1442472-39-0 | 98.7% | 510.36 | C24H21BrFN5O2 | 5 MG
SDP
Catalog No. 501872381
Encompass_Preferred
Ripretinib is a research-grade small-molecule kinase inhibitor that functions as a switch-control inhibitor of KIT and PDGFRA and displays activity against additional kinases. Supplied as a powdered reagent for laboratory research use only.
  • Targets wild-type and mutant KIT and PDGFRA.
  • Inhibits FLT3 and KDR/VEGFR-2.
  • Demonstrated oral bioavailability in preclinical studies.
  • High purity suitable for biochemical and cellular assays.
  • Supplied as a powder for flexible formulation and dosing.
  • Available in small research pack sizes for screening and pilot studies.
Catalog No. 50-187-2381 Supplier Medchemexpress LLC Supplier No. HY1123065MG
Ripretinib is a research-grade small-molecule kinase inhibitor that functions as a switch-control inhibitor of KIT and PDGFRA and displays activity against additional kinases. Supplied as a powdered reagent for laboratory research use only.
  • Targets wild-type and mutant KIT and PDGFRA.
  • Inhibits FLT3 and KDR/VEGFR-2.
  • Demonstrated oral bioavailability in preclinical studies.
  • High purity suitable for biochemical and cellular assays.
  • Supplied as a powder for flexible formulation and dosing.
  • Available in small research pack sizes for screening and pilot studies.