Learn More
Selleck Chemical LLC Ripretinib S8757-25MG
SDP
Catalog No. 5000024091
Encompass_Preferred
Ripretinib is an orally bioavailable switch pocket control inhibitor of wild-type and mutated forms of the tumor-associated antigens (TAA) mast/stem cell factor receptor (SCFR) Kit (c-Kit) and PDGFR-alpha with IC50 values of 4 nM 8 nM 18 nM 5 nM and 14 nM for WT Kit (c-Kit) V654A Kit (c-Kit) T670I Kit (c-Kit) D816H Kit (c-Kit) and D816V Kit (c-Kit) respectively
Catalog No. 50-000-24091 Supplier Selleck Chemical LLC Supplier No. S875725MG
Ripretinib is an orally bioavailable switch pocket control inhibitor of wild-type and mutated forms of the tumor-associated antigens (TAA) mast/stem cell factor receptor (SCFR) Kit (c-Kit) and PDGFR-alpha with IC50 values of 4 nM 8 nM 18 nM 5 nM and 14 nM for WT Kit (c-Kit) V654A Kit (c-Kit) T670I Kit (c-Kit) D816H Kit (c-Kit) and D816V Kit (c-Kit) respectively