Learn More
Selleck Chemical LLC Risankizumab-A2046-1MG
SDP
Supplier:  Selleck Chemical LLC A20461MG
Encompass_Preferred
Risankizumab targets the p19 subunit of IL-23 Risankizumab is indicated for the treatment of psoriasis and is safe and effective in maintaining remission in active Crohn s disease
Catalog No. 50-234-1171