Learn More
Medchemexpress LLC Risarestat | 79714-31-1 | 98.03% | 323.41 | 50 MG
SDP
Catalog No. 5000392441
Encompass_Preferred
Risarestat (CT-112) is an aldose reductase inhibitor developed for the treatment of diabetic complications. It is intended for research use only and not for sale to patients.
  • Aids in research of diabetic complications.
  • Acts as an aldose reductase inhibitor.
  • Supports biochemical investigations.
  • Useful for studying metabolic pathways.
  • Contributes to drug development efforts.
Catalog No. 50-003-92441 Supplier Medchemexpress LLC Supplier No. HY1643350MG
Risarestat (CT-112) is an aldose reductase inhibitor developed for the treatment of diabetic complications. It is intended for research use only and not for sale to patients.
  • Aids in research of diabetic complications.
  • Acts as an aldose reductase inhibitor.
  • Supports biochemical investigations.
  • Useful for studying metabolic pathways.
  • Contributes to drug development efforts.