Learn More
Medchemexpress LLC Risarestat (CT 112) | 79714-31-1 | 98.0% | 323.41 | 100 MG
SDP
Catalog No. 5000392350
Encompass_Preferred
Risarestat (CT-112) is an aldose reductase inhibitor developed for the treatment of diabetic complications. It inhibits the accumulation of dulcitol in a dose-dependent manner and shows intraocular penetration following topical instillation.
  • Inhibits accumulation of dulcitol
  • Shows intraocular penetration
  • Significantly decreases the anterior surface area of superficial corneal cells
  • Improves corneal sensitivity in diabetic patients
  • Increases blink response in galactose-fed rats
  • Solid with a white to light yellow appearance
Catalog No. 50-003-92350 Supplier Medchemexpress LLC Supplier No. HY16433100MG
Risarestat (CT-112) is an aldose reductase inhibitor developed for the treatment of diabetic complications. It inhibits the accumulation of dulcitol in a dose-dependent manner and shows intraocular penetration following topical instillation.
  • Inhibits accumulation of dulcitol
  • Shows intraocular penetration
  • Significantly decreases the anterior surface area of superficial corneal cells
  • Improves corneal sensitivity in diabetic patients
  • Increases blink response in galactose-fed rats
  • Solid with a white to light yellow appearance