Learn More
Medchemexpress LLC HY-109101 10mg Medchemexpress, Risdiplam CAS:1825352-65-5 Purity:>98%
SDP
Catalog No. 501968680
Encompass_Preferred
Medchemexpress, HY-109101 10mg Risdiplam CAS:1825352-65-5 Risdiplam (RG7916; RO7034067) is an orally administered, centrally and peripherally distributed SMN2 pre-mRNA splicing modifier that increases survival motor neuron (SMN) protein levels. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8680 Supplier Medchemexpress LLC Supplier No. HY10910110MG
Medchemexpress, HY-109101 10mg Risdiplam CAS:1825352-65-5 Risdiplam (RG7916; RO7034067) is an orally administered, centrally and peripherally distributed SMN2 pre-mRNA splicing modifier that increases survival motor neuron (SMN) protein levels. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.