Learn More
Medchemexpress LLC Risdiplam | 1825352-65-5 | 99.0% | 401.46 | 1 MG
SDP
Catalog No. 5000345185
Encompass_Preferred
Risdiplam (RG7916) is an orally administered, centrally and peripherally distributed SMN2 pre-mRNA splicing modifier that increases survival motor neuron (SMN) protein levels. It modulates SMN2 pre-mRNA splicing towards the production of full-length SMN2 mRNA, thereby increasing functional SMN protein.
  • Orally administered and distributed centrally and peripherally
  • Modulates SMN2 pre-mRNA splicing
  • Increases survival motor neuron (SMN) protein levels
  • Leads to an increase in SMN2 full length transcript
  • Suitable for laboratory centrifugation and storage
Catalog No. 50-003-45185 Supplier Medchemexpress LLC Supplier No. HY109101100MG
Risdiplam (RG7916) is an orally administered, centrally and peripherally distributed SMN2 pre-mRNA splicing modifier that increases survival motor neuron (SMN) protein levels. It modulates SMN2 pre-mRNA splicing towards the production of full-length SMN2 mRNA, thereby increasing functional SMN protein.
  • Orally administered and distributed centrally and peripherally
  • Modulates SMN2 pre-mRNA splicing
  • Increases survival motor neuron (SMN) protein levels
  • Leads to an increase in SMN2 full length transcript
  • Suitable for laboratory centrifugation and storage