Learn More
Medchemexpress LLC Risdiplam | 1825352-65-5 | 99.6% | 401.46 | 50 MG
SDP
Catalog No. 5000345445
Encompass_Preferred
Risdiplam (RG7916) is an orally administered, centrally and peripherally distributed SMN2 pre-mRNA splicing modifier that increases survival motor neuron (SMN) protein levels. For research use only.
  • Orally administered
  • Centrally and peripherally distributed
  • SMN2 pre-mRNA splicing modifier
  • Increases survival motor neuron (SMN) protein levels
Catalog No. 50-003-45445 Supplier Medchemexpress LLC Supplier No. HY10910150MG
Risdiplam (RG7916) is an orally administered, centrally and peripherally distributed SMN2 pre-mRNA splicing modifier that increases survival motor neuron (SMN) protein levels. For research use only.
  • Orally administered
  • Centrally and peripherally distributed
  • SMN2 pre-mRNA splicing modifier
  • Increases survival motor neuron (SMN) protein levels