Learn More
Medchemexpress LLC Risdiplam-d4 | 1825352-65-5 | 98.9% | 405.49 | 100 MG
SDP
Catalog No. 5000344630
Encompass_Preferred
Risdiplam-d4 is a deuterium-labeled version of Risdiplam. Risdiplam (RG7916) is an orally administered SMN2 pre-mRNA splicing modifier that is distributed both centrally and peripherally, and it works by increasing survival motor neuron (SMN) protein levels.
  • This compound can be used as a tracer.
  • It can serve as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS.
  • Deuteration can influence the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-44630 Supplier Medchemexpress LLC Supplier No. HY109101S1MG
Risdiplam-d4 is a deuterium-labeled version of Risdiplam. Risdiplam (RG7916) is an orally administered SMN2 pre-mRNA splicing modifier that is distributed both centrally and peripherally, and it works by increasing survival motor neuron (SMN) protein levels.
  • This compound can be used as a tracer.
  • It can serve as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS.
  • Deuteration can influence the pharmacokinetic and metabolic profiles of drugs.