Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Risedronate Related Compound C Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111001894 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-1894 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-1894 Supplier MilliporeSigma Supelco Supplier No. PHR207750MG
Add to Cart
Add to Cart
Risedronate Related Compound C is an impurity of risedronate, belonging to the group of pyridinyl bisphosphonates, and is effectively used in the treatment of Paget′s disease of bone. It is also used for the treatment of skeletal disorders since it inhibits osteoclast-mediated bone resorption and modulates bone metabolism.
TRUSTED_SUSTAINABILITY

Specifications

CAS 75755-10-1
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C7H11 NO6P2
MDL Number MFCD26142887
Synonym 2-(3-Pyridinyl)ethylidene]bis(phosphonic acid); 2-(Pyridin-3-yl)ethane-1,1-diyl]bis(phosphonic acid)
Type Certified Reference Material
Quantity 50 mg
Formula Weight 267.11
Grade Certified Reference Material
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.