Learn More
Apexbio Technology LLC Risedronate Sodium 115436-72-1 1g
SDP
Catalog No. 5000093498
Encompass_Preferred
Risedronate Sodium (115436-72-1) is a small-molecule inhibitor targeting farnesyl diphosphate (FPP) synthase It is designed to block FPP synthase thereby interfering with the mevalonate pathway and decreasing downstream protein prenylation necessary for cellular proliferation and survival Risedronate Sodium exerts its biological activity primarily through inhibition of FPP synthase leading to antiproliferative and proapoptotic effects in multiple tumor cell lines Additionally it modulates bone metabolism by suppressing osteoclast-mediated bone resorption Based on these pharmacological properties Risedronate Sodium holds research potential in tumor cell biology apoptosis pathways osteoporosis and metabolic bone disease models
Catalog No. 50-000-93498 Supplier Apexbio Technology LLC Supplier No. A52931000
Risedronate Sodium (115436-72-1) is a small-molecule inhibitor targeting farnesyl diphosphate (FPP) synthase It is designed to block FPP synthase thereby interfering with the mevalonate pathway and decreasing downstream protein prenylation necessary for cellular proliferation and survival Risedronate Sodium exerts its biological activity primarily through inhibition of FPP synthase leading to antiproliferative and proapoptotic effects in multiple tumor cell lines Additionally it modulates bone metabolism by suppressing osteoclast-mediated bone resorption Based on these pharmacological properties Risedronate Sodium holds research potential in tumor cell biology apoptosis pathways osteoporosis and metabolic bone disease models