Learn More
Cayman Chemical Risedronate (sodium salt), Purity: ≥95%, CAS: 115436-72-1, 500 mg
SDP
Catalog No. 502983549
Encompass_Preferred
An aminobisphosphonate and antimalarial agent that inhibits bone resorption; inhibits growth of P. falciparum in vitro (IC50 = 20.3 µM) and P. berghei in vivo (25 mg/kg for one week); inhibits protein prenylation by preventing transfer of a farnesyl pyrophosphate group to parasite proteins
Catalog No. 50-298-3549 Supplier Cayman Chemical Supplier No. 20740500 MG
An aminobisphosphonate and antimalarial agent that inhibits bone resorption; inhibits growth of P. falciparum in vitro (IC50 = 20.3 µM) and P. berghei in vivo (25 mg/kg for one week); inhibits protein prenylation by preventing transfer of a farnesyl pyrophosphate group to parasite proteins