Learn More
Medchemexpress LLC Risedronic acid sodium | 115436-72-1 | 98.0% | 1 ML
SDP
Catalog No. 5000450732
Encompass_Preferred
Risedronic acid sodium is a bisphosphonate and a potent anti-resorption agent. It inhibits osteoclast-mediated bone resorption and influences bone metabolism. This compound suppresses osteoblast differentiation and induces caspase- and isoprenoid depletion-dependent apoptosis, and has shown activity against Plasmodium falciparum by interfering with farnesyl pyrophosphate group transfer to parasite proteins.
  • Potent anti-resorption agent
  • Inhibits osteoclast-mediated bone resorption
  • Suppresses osteoblast differentiation
  • Induces caspase- and isoprenoid depletion-dependent apoptosis
  • Active against Plasmodium falciparum
  • Prevents radiation-induced osteoporosis in mice
  • Inhibits parasitemia in infected mice
Catalog No. 50-004-50732 Supplier Medchemexpress LLC Supplier No. HYB011910MM/1ML
Risedronic acid sodium is a bisphosphonate and a potent anti-resorption agent. It inhibits osteoclast-mediated bone resorption and influences bone metabolism. This compound suppresses osteoblast differentiation and induces caspase- and isoprenoid depletion-dependent apoptosis, and has shown activity against Plasmodium falciparum by interfering with farnesyl pyrophosphate group transfer to parasite proteins.
  • Potent anti-resorption agent
  • Inhibits osteoclast-mediated bone resorption
  • Suppresses osteoblast differentiation
  • Induces caspase- and isoprenoid depletion-dependent apoptosis
  • Active against Plasmodium falciparum
  • Prevents radiation-induced osteoporosis in mice
  • Inhibits parasitemia in infected mice