Learn More
Medchemexpress LLC HY-106441A 5mg Medchemexpress, Rislenemdaz CAS:808732-98-1 Purity:>98%
SDP
Catalog No. 501968343
Encompass_Preferred
Medchemexpress, HY-106441A 5mg Rislenemdaz CAS:808732-98-1 Rislenemdaz (CERC-301) is an orally bioavailable and selective N-methyl-D-aspartate (NMDA) receptor subunit 2B (GluN2B) antagonist with Ki and IC 50 of 8.1 nM and 3.6 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8343 Supplier Medchemexpress LLC Supplier No. HY106441A5MG
Medchemexpress, HY-106441A 5mg Rislenemdaz CAS:808732-98-1 Rislenemdaz (CERC-301) is an orally bioavailable and selective N-methyl-D-aspartate (NMDA) receptor subunit 2B (GluN2B) antagonist with Ki and IC 50 of 8.1 nM and 3.6 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.