Learn More
AdipoGen Risperidone (50 mg)
SDP
Supplier:  AdipoGen CDXR0125M050
Encompass_Preferred
Chemical. CAS: 106266-06-2. Formula: C17H26N2O. MW: 410.48. Risperidone is a benzisoxazole derivative and serotonin-dopamine antagonist. It is an atypical antipsychotic that binds to dopamine D2 receptors (Ki=3nM) and the serotonin (5-HT) receptor subtype 5-HT2A (Ki=0.12nM). It also binds to dopamine D4, ?1- and ?2-adrenergic, 5-HT1C, 5-HT1D, and histamine H1 receptors. It therefore functions as an antagonist for dopamine D2, histamine H1 and alpha1 and alpha2 adrenergic receptors. Formulations containing risperidone have been used in the treatment of schizophrenia, autism and bipolar disorder.
Catalog No. 50-205-4633