Learn More
Medchemexpress LLC Risperidone-d4 | 1020719-76-9 | 99.4% | 414.51 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1102321MG
Encompass_Preferred
Risperidone-d4 is the deuterium labeled Risperidone. Risperidone is a serotonin 5-HT2 receptor blocker, P-Glycoprotein inhibitor and potent dopamine D2 receptor antagonist, with Kis of 4.8, 5.9 nM for 5-HT2A and dopamine D2 receptor, respectively. This compound is for research use only and not sold to patients. This compound can be used as a tracer or as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-44754