Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Risperidone Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111018708 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8708 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8708 Supplier MilliporeSigma Supelco Supplier No. PHR1631500MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Risperidone is a new central 5-hydroxytryptamine2 and dopamine D2 antagonist. It is used to treat agitation, aggression,and psychosis caused during demnetia. It is a neuroleptic drug. It is also an atypical antipsychotic drug which is used for treating schizophrenia.

TRUSTED_SUSTAINABILITY

Specifications

CAS 106266-06-2
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C23H27 FN4O2
MDL Number MFCD00274576
Synonym Risperidone; R 64,766
Type Certified Reference Material
Quantity 500 mg
Formula Weight 410.48
Grade Certified Reference Material
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.