Learn More
Selleck Chemical LLC Risperidone 10mg 106266-06-2 R-64766
SDP
Supplier:  Selleck Chemical LLC S161510MG
Encompass_Preferred
Risperidone (R-64766) is a mutil-targeted antagonist for dopamine, serotonin, adrenergic and histamine receptors, used to treat schizophrenia and bipolar disorder. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-588-5