Learn More
Cayman Chemical Risperidone-d4, Purity: ≥99% deuterated forms (d1-d4), CAS: 1020719-76-9, 1 mg
SDP
Catalog No. 502887818
Encompass_Preferred
An internal standard for the quantification of risperidone by GC- or LC-MS
Catalog No. 50-288-7818 Supplier Cayman Chemical Supplier No. 100105701 MG
An internal standard for the quantification of risperidone by GC- or LC-MS