Learn More
Cayman Chemical Risperidoned4 GC MS Standards
SDP
Supplier:  Cayman Chemical 10010570500 UG
Encompass_Preferred
An internal standard for the quantification of risperidone by GC- or LC-MS
Catalog No. 50-288-7784