Learn More
Enzo Life Sciences Ristocetin A. sulfate (10mg). CAS: 11140-99-1
SDP
Supplier:  Enzo Life Sciences ALX380237M010
Encompass_Preferred
Potent antibacterial antibiotic. Induces platelet aggregation by binding to a factor absent in people suffering from von Willebrand’s disease and is therefore an important diagnostic tool. Chiral selector for the separation of enantiomers by HPLC, TLC and electrophoresis. Alternative name: Ristocetin . sulfate, Ristomycin III . sulfate, Spontin . sulfate, Riston . sulfate. Purity: ≥95% (HPLC). Solubility: Soluble in water, methanol or DMSO. Long Term Storage: -20°C.
Catalog No. 50-200-8985