Learn More
Cayman Chemical RIstocetIn Asulfate 10mg
SDP
Catalog No. 502982963
Encompass_Preferred
An antibiotic used in the ristocetin cofactor assay and the ristocetin-induced platelet aggregation assay, for the diagnosis of von Willebrand disease and other bleeding disorders
Catalog No. 50-298-2963 Supplier Cayman Chemical Supplier No. 2060110 MG
An antibiotic used in the ristocetin cofactor assay and the ristocetin-induced platelet aggregation assay, for the diagnosis of von Willebrand disease and other bleeding disorders