Learn More
Cayman Chemical Ristocetin A (sulfate), Purity: ≥85%, CAS: 92989-99-6, 50 mg
SDP
Catalog No. 502984500
Encompass_Preferred
An antibiotic used in the ristocetin cofactor assay and the ristocetin-induced platelet aggregation assay, for the diagnosis of von Willebrand disease and other bleeding disorders
Catalog No. 50-298-4500 Supplier Cayman Chemical Supplier No. 2060150 MG
An antibiotic used in the ristocetin cofactor assay and the ristocetin-induced platelet aggregation assay, for the diagnosis of von Willebrand disease and other bleeding disorders