Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ristocetin Sulfate Salt, >90% Ristocetin A; remainder mostly Ristocetin B., (Crystalline), MP Biomedicals

Catalog No. ICN19548905 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19548905 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19548905 Supplier MP Biomedicals, Inc Supplier No. 0219548905
Add to Cart
Add to Cart

Useful for investigation of platelet aggregation

TRUSTED_SUSTAINABILITY

Specifications

Assay >90% Ristocetin A; remainder mostly Ristocetin B.
CAS 1404-55-3
Content And Storage 0 to 5°C
Format Crystalline
Molecular Weight (g/mol) 2166.009
Product Type Ristocetin Sulfate Salt
Quantity 5 mg
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.