Learn More
Sigma Aldrich Fine Chemicals Biosciences Ristomycin monosulfate 90100MG
Catalog No. 502903172
Encompass
Ristomycin/ristocetin is a type III glycopeptide. It is a highly glycosylated glycopeptide previously found innbspAmycolatopsis lurida.
Catalog No. 50-290-3172 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. R7752100MG
Ristomycin/ristocetin is a type III glycopeptide. It is a highly glycosylated glycopeptide previously found innbspAmycolatopsis lurida.