Learn More
Medchemexpress LLC HY-13424 5mg Medchemexpress, RITA CAS:213261-59-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY134245MG
Encompass_Preferred
Medchemexpress, HY-13424 5mg RITA CAS:213261-59-7 RITA is an inhibitor of p53-HDM-2 interaction, binds to p53dN, with a Kd of 1.5 nM, and also induces DNA-DNA cross-links. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0309