Learn More
Abcam RITA, p53 activator, 5MG
Catalog No. 502672808
Encompass_Preferred
MW 292.4 Da, Purity >98%. p53 activator. Tricyclic thiophene derivative that binds to p53 and induces its accumulation in tumor cells. Prevents p53-HDM2 (MDM2) interaction in vitro and in vivo and affects p53 interaction with several negative regulators. Induces expression of p53 target genes and apoptosis in various tumor cell lines expressing wild-type p53.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-2808 Supplier Abcam Supplier No. AB2193795MG
MW 292.4 Da, Purity >98%. p53 activator. Tricyclic thiophene derivative that binds to p53 and induces its accumulation in tumor cells. Prevents p53-HDM2 (MDM2) interaction in vitro and in vivo and affects p53 interaction with several negative regulators. Induces expression of p53 target genes and apoptosis in various tumor cell lines expressing wild-type p53.
The product is subject to the following: Abcam Restricted Use Statement