Learn More
Abcam RITA, p53 activator, 25MG
Catalog No. 502672809
Encompass_Preferred
MW 292.4 Da, Purity >98%. p53 activator. Tricyclic thiophene derivative that binds to p53 and induces its accumulation in tumor cells. Prevents p53-HDM2 (MDM2) interaction in vitro and in vivo and affects p53 interaction with several negative regulators. Induces expression of p53 target genes and apoptosis in various tumor cell lines expressing wild-type p53.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-2809 Supplier Abcam Supplier No. AB21937925MG
MW 292.4 Da, Purity >98%. p53 activator. Tricyclic thiophene derivative that binds to p53 and induces its accumulation in tumor cells. Prevents p53-HDM2 (MDM2) interaction in vitro and in vivo and affects p53 interaction with several negative regulators. Induces expression of p53 target genes and apoptosis in various tumor cell lines expressing wild-type p53.
The product is subject to the following: Abcam Restricted Use Statement