Learn More
Medchemexpress LLC Ritalinic acid | 19395-41-6 | MFCD06200695 | 99.8% | 219.28 g/mol | C13H17NO2 | 250 MG
SDP
Catalog No. 5000208648
Encompass_Preferred
Ritalinic acid is the primary inactive urinary metabolite of methylphenidate, supplied as a powder for research use. It is a small organic acid (CAS 19395-41-6, C13H17NO2, MW 219.28 g/mol) provided at high purity for biochemical and pharmacological studies related to attention-deficit hyperactivity disorder and narcolepsy research.
  • High purity (99.81%) suitable for analytical and research applications.
  • Available in multiple package sizes including 250 MG powder.
  • Stable as powder: -20°C (3 years), 4°C (2 years).
  • Solvent storage stability: -80°C (6 months), -20°C (1 month).
Catalog No. 50-002-08648 Supplier Medchemexpress LLC Supplier No. 250MGHYW042214
Ritalinic acid is the primary inactive urinary metabolite of methylphenidate, supplied as a powder for research use. It is a small organic acid (CAS 19395-41-6, C13H17NO2, MW 219.28 g/mol) provided at high purity for biochemical and pharmacological studies related to attention-deficit hyperactivity disorder and narcolepsy research.
  • High purity (99.81%) suitable for analytical and research applications.
  • Available in multiple package sizes including 250 MG powder.
  • Stable as powder: -20°C (3 years), 4°C (2 years).
  • Solvent storage stability: -80°C (6 months), -20°C (1 month).