Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ritalinic acid | 19395-41-6 | MFCD06200695 | 99.8% | 219.28 g/mol | C13H17NO2 | 5 MG
SDP

Catalog No. 5000209260
Encompass_Preferred

Ritalinic acid is the primary inactive urinary metabolite of methylphenidate, supplied as a high-purity research reagent for metabolite, pharmacokinetic, and analytical studies. It is intended for research use only and not for human or clinical applications.

  • Purity 99.8%.
  • CAS number 19395-41-6.
  • Molecular formula C13H17NO2.
  • Molecular weight 219.28 g/mol.
  • Available in small pack sizes, including 5 mg.
  • Intended use: research reagent for metabolite and analytical studies.

Catalog No. 50-002-09260 Supplier Medchemexpress LLC Supplier No. HYW0422145MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Ritalinic acid is the primary inactive urinary metabolite of methylphenidate, supplied as a high-purity research reagent for metabolite, pharmacokinetic, and analytical studies. It is intended for research use only and not for human or clinical applications.

  • Purity 99.8%.
  • CAS number 19395-41-6.
  • Molecular formula C13H17NO2.
  • Molecular weight 219.28 g/mol.
  • Available in small pack sizes, including 5 mg.
  • Intended use: research reagent for metabolite and analytical studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.