Learn More
Cayman Chemical Ritalinic Acid, Purity: ≥98%, CAS: 19395-41-6, 50 mg
SDP
Catalog No. 502973174
Encompass_Preferred
An inactive, major metabolite of methylphenidate; intended for forensic and biological research purposes
Catalog No. 50-297-3174 Supplier Cayman Chemical Supplier No. 1591550 MG
An inactive, major metabolite of methylphenidate; intended for forensic and biological research purposes