Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ritlecitinib (PF-06651600) | 1792180-81-4 | MFCD30343868 | 99.8% | 285.35 g/mol | C15H19N5O | 10 MG
SDP

Catalog No. 5000212139
Encompass_Preferred

Ritlecitinib is a selective JAK3 inhibitor research reagent supplied as a 10 mg solid for in vitro and preclinical studies. It is the (2R,5S) stereoisomer and is intended for research use only, not for human or therapeutic use.

  • High purity (99.8%).
  • Molecular weight 285.35 g/mol.
  • Chemical formula C15H19N5O.
  • CAS number 1792180-81-4.
  • Selective JAK3 inhibitor with reported IC50 ≈ 33 nM.
  • Supplied as a 10 mg solid for laboratory research.
  • Intended for in vitro and preclinical research; not for therapeutic use.

Catalog No. 50-002-12139 Supplier Medchemexpress LLC Supplier No. HY100754B10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ritlecitinib is a selective JAK3 inhibitor research reagent supplied as a 10 mg solid for in vitro and preclinical studies. It is the (2R,5S) stereoisomer and is intended for research use only, not for human or therapeutic use.

  • High purity (99.8%).
  • Molecular weight 285.35 g/mol.
  • Chemical formula C15H19N5O.
  • CAS number 1792180-81-4.
  • Selective JAK3 inhibitor with reported IC50 ≈ 33 nM.
  • Supplied as a 10 mg solid for laboratory research.
  • Intended for in vitro and preclinical research; not for therapeutic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.