Learn More
Apexbio Technology LLC Ritodrine HCl 23239-51-2 100mg
SDP
Catalog No. 502240810
Encompass_Preferred
Ritodrine hydrochloride (CAS 23239-51-2) is a selective 2-adrenergic receptor agonist that modulates smooth muscle activity By stimulating 2-adrenergic receptors ritodrine promotes uterine muscle relaxation thereby inhibiting premature uterine contractions Comparative studies indicate that ritodrine can rapidly extend the interval before delivery although it is associated with a higher incidence of adverse effects compared to alternatives such as magnesium sulfate Ritodrine HCl is widely used in research to investigate 2-adrenergic signaling pathways and to model the pharmacological regulation of preterm labor
This item is not returnable. View return policy
Catalog No. 50-224-0810 Supplier Apexbio Technology LLC Supplier No. B1347100
Ritodrine hydrochloride (CAS 23239-51-2) is a selective 2-adrenergic receptor agonist that modulates smooth muscle activity By stimulating 2-adrenergic receptors ritodrine promotes uterine muscle relaxation thereby inhibiting premature uterine contractions Comparative studies indicate that ritodrine can rapidly extend the interval before delivery although it is associated with a higher incidence of adverse effects compared to alternatives such as magnesium sulfate Ritodrine HCl is widely used in research to investigate 2-adrenergic signaling pathways and to model the pharmacological regulation of preterm labor