Learn More
Medchemexpress LLC Ritodrine hydrochloride | 23239-51-2 | 98.5% | 100 MG
SDP
Catalog No. 5000408958
Encompass_Preferred
Ritodrine hydrochloride is a potent and orally active β-adrenergic agonist. It is known to decrease preterm delivery and uterine contraction, showing potential for research into preterm labor.
  • Potent and orally active beta-adrenergic agonist.
  • Decreases preterm delivery.
  • Decreases uterine contraction.
  • Potential for research of preterm labor.
  • Targets beta adrenergic receptor.
Catalog No. 50-004-08958 Supplier Medchemexpress LLC Supplier No. HYB0452100MG
Ritodrine hydrochloride is a potent and orally active β-adrenergic agonist. It is known to decrease preterm delivery and uterine contraction, showing potential for research into preterm labor.
  • Potent and orally active beta-adrenergic agonist.
  • Decreases preterm delivery.
  • Decreases uterine contraction.
  • Potential for research of preterm labor.
  • Targets beta adrenergic receptor.