Learn More
Medchemexpress LLC Ritodrine hydrochloride | 23239-51-2 | 98.5% | 500 MG
SDP
Catalog No. 5000407760
Encompass_Preferred
Ritodrine (DU21220) hydrochloride is a potent and orally active β-adrenergic agonist. It decreases preterm delivery and uterine contraction, showing potential for research into preterm labor.
  • Potent and orally active β-adrenergic agonist
  • Decreases preterm delivery and uterine contraction
  • Useful in research for preterm labor
Catalog No. 50-004-07760 Supplier Medchemexpress LLC Supplier No. HYB0452500MG
Ritodrine (DU21220) hydrochloride is a potent and orally active β-adrenergic agonist. It decreases preterm delivery and uterine contraction, showing potential for research into preterm labor.
  • Potent and orally active β-adrenergic agonist
  • Decreases preterm delivery and uterine contraction
  • Useful in research for preterm labor