Learn More
Cayman Chemical Ritodrine (hydrochloride), Purity: ≥98%, CAS: 23239-51-2, 1 g
SDP
Catalog No. 502936151
Encompass_Preferred
A β2-AR agonist; induces relaxation of isolated pig detrusor muscle from 0.001-1 μM; reduces electrically-induced isometric tension in myometrial strips isolated from rabbits on day 25 of pregnancy at 30 μg/ml; suppresses spontaneous motility of and increases concentration of cAMP in isolated pregnant rat uterus
Catalog No. 50-293-6151 Supplier Cayman Chemical Supplier No. 260841 G
A β2-AR agonist; induces relaxation of isolated pig detrusor muscle from 0.001-1 μM; reduces electrically-induced isometric tension in myometrial strips isolated from rabbits on day 25 of pregnancy at 30 μg/ml; suppresses spontaneous motility of and increases concentration of cAMP in isolated pregnant rat uterus