Learn More
Medchemexpress LLC HY-90001 500mg , Ritonavir A 84538;ABT 538;Abbott 84538;NSC 693184;RTV CAS:155213-67-5 Purity:98%
SDP
Catalog No. 501492383
Encompass_Preferred
Medchemexpress, HY-90001 500mg Ritonavir A 84538;ABT 538;Abbott 84538;NSC 693184;RTV CAS:155213-67-5 Formula:C37H48N6O5S2 Purity:98% Ritonavir is an inhibitor of HIV protease used to treat HIV infection and AIDS. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2383 Supplier Medchemexpress LLC Supplier No. HY90001500MG
Medchemexpress, HY-90001 500mg Ritonavir A 84538;ABT 538;Abbott 84538;NSC 693184;RTV CAS:155213-67-5 Formula:C37H48N6O5S2 Purity:98% Ritonavir is an inhibitor of HIV protease used to treat HIV infection and AIDS. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.