Learn More
Medchemexpress LLC Ritonavir-d6 | 1616968-73-0 | 98.8% | C37H42D6N6O5S2 | 1 MG
SDP
Catalog No. 5000433191
Encompass_Preferred
Ritonavir-d6 is the deuterium-labeled form of Ritonavir, an inhibitor of HIV protease used in the treatment of HIV infection and AIDS. It also functions as a SARS-CoV 3CLpro inhibitor with an IC50 of 1.61 μM. This compound can be used as a tracer and as an internal standard for quantitative analysis. Deuterium substitution is used to study the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Chemical reagent for research use only
  • Not intended for human use
Catalog No. 50-004-33191 Supplier Medchemexpress LLC Supplier No. HY90001S1MG
Ritonavir-d6 is the deuterium-labeled form of Ritonavir, an inhibitor of HIV protease used in the treatment of HIV infection and AIDS. It also functions as a SARS-CoV 3CLpro inhibitor with an IC50 of 1.61 μM. This compound can be used as a tracer and as an internal standard for quantitative analysis. Deuterium substitution is used to study the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Chemical reagent for research use only
  • Not intended for human use