Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ritter Pentafluoroiodoethane-Dmso Reagent

Catalog No. 7944815ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
79-448-15ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 79-448-15ML Supplier Sigma Aldrich Supplier No. 7944815ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Product is a more convenient, liquified version of pentafluoroiodoethane, which can be utilized in photocatalytic and electrophilic pentafluoroethylation of olefins and heteroatoms respectively.Condensed-Phase, Halogen-Bonded CF3I and C2F5I Adducts for Perfluoroalkylation Reactions

Specifications

CAS 140-07-8
Density 1.1 g/mL at 20°C
Boiling Point ∼280°C
Molecular Formula C10H24N2O4
Linear Formula (HOCH2CH2)2NCH2CH2N(CH2CH2OH)2
Quantity 5 mL
MDL Number MFCD00002854
Synonym 2DMSO-CF3CF2I
Molecular Weight (g/mol) 236.31 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.