Learn More
TARGETMOL CHEMICALS INC RITUXIMAB 5MG
SDP
Catalog No. 502578933
Encompass_Preferred
Also available in 1 mg 10 mg and bulk. Please contact Fisher for quotes. Rituximab anti-CD20) is a chimeric anti-CD20 mAb that binds the CD20 antigen on B cells with a binding affinity of 5 nM. purity: 98%
Catalog No. 50-257-8933 Supplier TARGETMOL CHEMICALS INC Supplier No. T99105MG
Also available in 1 mg 10 mg and bulk. Please contact Fisher for quotes. Rituximab anti-CD20) is a chimeric anti-CD20 mAb that binds the CD20 antigen on B cells with a binding affinity of 5 nM. purity: 98%