Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TechNiGlove International Rival 12 in. Cl10 Nitrile Glove
SDP

Catalog No. 17998055 Shop All TechNiGlove International Inc. Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-998-055 2X-Large
17-998-050 X-Small
17-998-051 Small
17-998-052 Medium
17-998-053 Large
17-998-054 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 17-998-055 Supplier TechNiGlove International Supplier No. RVCR3505
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Accelerator Free Powder Free Soft Modulus 12 in. 100% Nitrile Class 10 Cleanroom Glove

  • Developed with TechNiGlove International's Soft Modulus Nitrile technology, providing a perfect balance of comfort, strength, and tactile sensitivity.
  • Accelerator-free and powder-free design minimizes skin irritation while maintaining a contaminant-free surface ideal for sensitive applications.
  • Constructed from 100% nitrile material for superior chemical resistance and reliable barrier protection in cleanroom conditions.
  • Offers particle counts as low as 250 particles per square centimeter-making it one of the cleanest ISO4 gloves available in the industry.
  • Delivers up to 220% elongation before breaking, reducing hand fatigue by over 50% compared to conventional nitrile gloves.
  • Extended 12-inch length ensures enhanced coverage and protection for wrists and forearms during precision work.
  • Perfect for semiconductor manufacturing, electronics production, battery assembly, and other controlled ISO4 environments.

Specifications

Chlorinated Yes
Cleanroom Class Class 1 (ISO 3)
Accelerator Free Yes
AQL 1.5
Color White
Disposable Single-use
Exterior Finish Fingertip Textured
Finger Thickness (English) 4 mil
Finger Thickness (Metric) 0.10 mm
Hand Ambidextrous
Latex Free Yes
Material Nitrile
Palm Thickness (English) 4 mil
Palm Thickness (Metric) 0.10 mm
Powdered Powder Free
Product Type Controlled Environment Gloves
Size 2X-Large
Sterility Non-sterile
Thickness (English) 4 mil
Thickness (Metric) 0.10 mm
Packaging Type Bag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.