Learn More
TARGETMOL CHEMICALS INC RIVAROXABAN 50MG
SDP
Catalog No. 502577442
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 25 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Rivaroxaban (BAY 59-7939) is an orally bioavailable oxazolidinone derivative and direct inhibitor of the coagulation factor Xa with anticoagulant activity. Upon oral administration rivaroxaban selectively binds to both free factor Xa and factor Xa bound in the prothrombinase complex. This interferes with the conversion of prothrombin (factor II) to thrombin and eventually prevents the formation of cross-linked fibrin clots. Rivaroxaban does not affect existing thrombin levels. purity: 99%
Catalog No. 50-257-7442 Supplier TARGETMOL CHEMICALS INC Supplier No. T118450MG
Also available in 2 mg 5 mg 10 mg 25 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Rivaroxaban (BAY 59-7939) is an orally bioavailable oxazolidinone derivative and direct inhibitor of the coagulation factor Xa with anticoagulant activity. Upon oral administration rivaroxaban selectively binds to both free factor Xa and factor Xa bound in the prothrombinase complex. This interferes with the conversion of prothrombin (factor II) to thrombin and eventually prevents the formation of cross-linked fibrin clots. Rivaroxaban does not affect existing thrombin levels. purity: 99%