Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rivaroxaban, 98%

Catalog No. AC461600010
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC461600010 1 g
AC461602500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC461600010 Supplier Thermo Scientific Chemicals Supplier No. 461600010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 366789-02-8 | C19H18ClN3O5S | 435.88 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 366789-02-8
Molecular Formula C19H18ClN3O5S
Molecular Weight (g/mol) 435.88
MDL Number MFCD11974010
InChI Key KGFYHTZWPPHNLQ-UHFFFAOYNA-N
Synonym rivaroxaban, xarelto, s-5-chloro-n-2-oxo-3-4-3-oxomorpholino phenyl oxazolidin-5-yl methyl thiophene-2-carboxamide, unii-9ndf7jz4m3, xarelto tn, 9ndf7jz4m3, 5-chloro-n-5s-2-oxo-3-4-3-oxomorpholin-4-yl phenyl-1,3-oxazolidin-5-yl methyl thiophene-2-carboxamide, 2-thiophenecarboxamide, 5-chloro-n-5s-2-oxo-3-4-3-oxo-4-morpholinyl phenyl-5-oxazolidinyl methyl, 5-chloro-n-5s-2-oxo-3-4-3-oxo-4-morpholinyl phenyl-5-oxazolidinyl methyl-2-thiophenecarboxamide
PubChem CID 9875401
ChEBI CHEBI:68579
IUPAC Name 5-chloro-N-[[(5S)-2-oxo-3-[4-(3-oxomorpholin-4-yl)phenyl]-1,3-oxazolidin-5-yl]methyl]thiophene-2-carboxamide
SMILES ClC1=CC=C(S1)C(=O)NCC1CN(C(=O)O1)C1=CC=C(C=C1)N1CCOCC1=O

Specifications

Melting Point 228.0°C to 229.0°C
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 435.88
Percent Purity 98%
Chemical Name or Material Rivaroxaban
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Call a POISON CENTER or doctor/physician if you feel unwell. Wash face, hands and any exposed skin thoroughly after handling. Avoid release to the environment. Collect spillage. Dispose of contents/container to an approved waste disposal plant.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.