Learn More
Selleck Chemical LLC Rivaroxaban 50mg 366789-02-8 BAY 59-7939
SDP
Supplier:  Selleck Chemical LLC S300250MG
Encompass_Preferred
Rivaroxaban (BAY 59-7939) is a direct inhibitor of Factor Xa with Ki and IC50 of 0.4 nM and 0.7 nM in cell-free assays, respectively. It is selective for human factor Xa, for which it has >10 000-fold greater selectivity than for other biologically relevant serine proteases (IC50 >20 ?M). *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-618-8