Learn More
Medchemexpress LLC Rivaroxaban-d4 (BAY 59-7939-d4) | 1132681-38-9 | 98.7% | 439.91 | 1 MG
SDP
Catalog No. 5000365482
Encompass_Preferred
Rivaroxaban-d4 (BAY 59-7939-d4) is a deuterium-labeled form of Rivaroxaban, a highly potent, selective, and direct Factor Xa (FXa) inhibitor. It demonstrates significant anti-FXa potency with an IC50 of 0.7 nM and a Ki of 0.4 nM.
  • Deuterium labeled compound
  • Potent, selective, and direct Factor Xa (FXa) inhibitor
  • Useful as a tracer or internal standard
  • Suitable for quantitative analysis by NMR, GC-MS, or LC-MS
  • For research use only
Catalog No. 50-003-65482 Supplier Medchemexpress LLC Supplier No. 1MGHY50903S
Rivaroxaban-d4 (BAY 59-7939-d4) is a deuterium-labeled form of Rivaroxaban, a highly potent, selective, and direct Factor Xa (FXa) inhibitor. It demonstrates significant anti-FXa potency with an IC50 of 0.7 nM and a Ki of 0.4 nM.
  • Deuterium labeled compound
  • Potent, selective, and direct Factor Xa (FXa) inhibitor
  • Useful as a tracer or internal standard
  • Suitable for quantitative analysis by NMR, GC-MS, or LC-MS
  • For research use only